MilliporeSigma
  • Development of novel EDG3 antagonists using a 3D database search and their structure-activity relationships.
Development of novel EDG3 antagonists using a 3D database search and their structure-activity relationships.
Journal of medicinal chemistry (2002-10-04)
Yuuki Koide, Takeshi Hasegawa, Atsuo Takahashi, Akira Endo, Naoki Mochizuki, Masako Nakagawa, Atsushi Nishida
ABSTRACT
Sphingosine-1-phosphate (S1P) is an intracellular second messenger and an extracellular mediator through endothelial differentiation gene (EDG) receptors, which are a novel class of G-protein-coupled receptors. Although EDG has attracted much attention because of its various roles, no selective agonists or antagonists have yet been developed. This could account for the delay in clarifying the physiological roles of members of the EDG family. Because precise structural information on EDG receptors is not yet available, pharmacophore models were generated based on structural information for S1P using the rational drug design software Catalyst. Novel antagonists, 2-alkylthiazolidine-4-carboxylic acids, were retrieved from a three-dimensional database search using the pharmacophore models, and these showed activity for EDG3. On the basis of their nonphosphoric acid structure, more potent antagonists, 2-(m- or p-heptylphenyl)thiazolidine-4-carboxylic acid, were developed.
Sigma-Aldrich
CAY10444, ≥98% (HPLC)
Pricing and availability is not currently available.