MilliporeSigma
  • Analysis of conditional colocalization relationships and hierarchies in three-color microscopy images.
Analysis of conditional colocalization relationships and hierarchies in three-color microscopy images.
The Journal of cell biology (2022-05-14)
Jesus Vega-Lugo, Bruno da Rocha-Azevedo, Aparajita Dasgupta, Khuloud Jaqaman
ABSTRACT
Colocalization analysis of multicolor microscopy images is a cornerstone approach in cell biology. It provides information on the localization of molecules within subcellular compartments and allows the interrogation of known molecular interactions in their cellular context. However, almost all colocalization analyses are designed for two-color images, limiting the type of information that they reveal. Here, we describe an approach, termed "conditional colocalization analysis," for analyzing the colocalization relationships between three molecular entities in three-color microscopy images. Going beyond the question of whether colocalization is present or not, it addresses the question of whether the colocalization between two entities is influenced, positively or negatively, by their colocalization with a third entity. We benchmark the approach and showcase its application to investigate receptor-downstream adaptor colocalization relationships in the context of functionally relevant plasma membrane locations. The software for conditional colocalization analysis is available at https://github.com/kjaqaman/conditionalColoc.
Sigma-Aldrich
Anti-Integrin β1 Antibody, clone P4G11, clone P4G11, Chemicon®, from mouse
Pricing and availability is not currently available.