MilliporeSigma
  • Assembling stable syntrophic Escherichia coli communities by comprehensively identifying beneficiaries of secreted goods.
Assembling stable syntrophic Escherichia coli communities by comprehensively identifying beneficiaries of secreted goods.
Cell systems (2021-09-02)
Mariana Noto Guillen, Brittany Rosener, Serkan Sayin, Amir Mitchell
ABSTRACT
Metabolic cross-feeding frequently underlies mutualistic relationships in natural microbial communities and is often exploited to assemble synthetic microbial consortia. We systematically identified all single-gene knockouts suitable for imposing cross-feeding in Escherichia coli and used this information to assemble syntrophic communities. Most strains benefiting from shared goods were dysfunctional in biosynthesis of amino acids, nucleotides, and vitamins or mutants in central carbon metabolism. We tested cross-feeding potency in 1,444 strain pairs and mapped the interaction network between all functional groups of mutants. This network revealed that auxotrophs for vitamins are optimal cooperators. Lastly, we monitored how assemblies composed of dozens of auxotrophs change over time and observed that they rapidly and repeatedly coalesced to seven strain consortia composed primarily from vitamin auxotrophs. The composition of emerging consortia suggests that they were stabilized by multiple cross-feeding interactions. We conclude that vitamins are ideal shared goods since they optimize consortium growth while still imposing member co-dependence.