MilliporeSigma
  • Opening Magnesium Storage Capability of Two-Dimensional MXene by Intercalation of Cationic Surfactant.
Opening Magnesium Storage Capability of Two-Dimensional MXene by Intercalation of Cationic Surfactant.
ACS nano (2018-03-16)
Min Xu, Shulai Lei, Jing Qi, Qingyun Dou, Lingyang Liu, Yulan Lu, Qing Huang, Siqi Shi, Xingbin Yan
ABSTRACT
Two-dimensional (2D) Ti3C2 MXene has attracted great attention in electrochemical energy storage devices (supercapacitors and lithium-ion and sodium-ion batteries) due to its excellent electrical conductivity as well as high volumetric capacity. Nevertheless, a previous study showed that multivalent Mg2+ ions cannot reversibly insert into MXene, resulting in a negligible capacity. Here, we demonstrate a simple strategy to achieve high magnesium storage capability for Ti3C2 MXene by preintercalating a cationic surfactant, cetyltrimethylammonium bromide (CTAB). Density functional theory simulations verify that intercalated CTA+ cations reduce the diffusion barrier of Mg2+ on the MXene surface, resulting in the significant improvement of the reversible insertion/deinsertion of Mg2+ ions between MXene layers. Consequently, the MXene electrode exhibits a desirable volumetric specific capacity of 300 mAh cm-3 at 50 mA g-1 as well as outstanding rate performance. This work endows MXene material with an application in electrochemical energy storage and, simultaneously, introduces magnesium battery materials as a member.
Sigma-Aldrich
Hydrophobic MXene for composites