MilliporeSigma
  • Ras-Related Protein Rab-32 and Thrombospondin 1 Confer Resistance to the EGFR Tyrosine Kinase Inhibitor Osimertinib by Activating Focal Adhesion Kinase in Non-Small Cell Lung Cancer.
Ras-Related Protein Rab-32 and Thrombospondin 1 Confer Resistance to the EGFR Tyrosine Kinase Inhibitor Osimertinib by Activating Focal Adhesion Kinase in Non-Small Cell Lung Cancer.
Cancers (2022-07-28)
Zeinab Kosibaty, Odd Terje Brustugun, Inger Johanne Zwicky Eide, Georgios Tsakonas, Oscar Grundberg, Luigi De Petris, Marc McGowan, Per Hydbring, Simon Ekman
ABSTRACT
Treatment with the tyrosine kinase inhibitor (TKI) osimertinib is the standard of care for non-small cell lung cancer (NSCLC) patients with activating mutations in the epidermal growth factor receptor (EGFR). Osimertinib is also used in T790M-positive NSCLC that may occur de novo or be acquired following first-line treatment with other EGFR TKIs (i.e., gefitinib, erlotinib, afatinib, or dacomitinib). However, patients treated with osimertinib have a high risk of developing resistance to the treatment. A substantial fraction of the mechanisms for resistance is unknown and may involve RNA and/or protein alterations. In this study, we investigated the full transcriptome of parental and osimertinib-resistant cell lines, revealing 131 differentially expressed genes. Knockdown screening of the genes upregulated in resistant cell lines uncovered eight genes to partly confer resistance to osimertinib. Among them, we detected the expression of Ras-related protein Rab-32 (RAB32) and thrombospondin 1 (THBS1) in plasmas sampled at baseline and at disease progression from EGFR-positive NSCLC patients treated with osimertinib. Both genes were upregulated in progression samples. Moreover, we found that knockdown of RAB32 and THBS1 reduced the expression of phosphorylated focal adhesion kinase (FAK). Combination of osimertinib with a FAK inhibitor resulted in synergistic toxicity in osimertinib-resistant cells, suggesting a potential therapeutic drug combination for overcoming resistance to osimertinib in NSCLC patients.
Sigma-Aldrich
Anti-RAB32 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution