Merck
  • Utility of Phox2b immunohistochemical stain in neural crest tumours and non-neural crest tumours in paediatric patients.
Utility of Phox2b immunohistochemical stain in neural crest tumours and non-neural crest tumours in paediatric patients.
Histopathology (2017-10-08)
Mikako Warren, Ryosuke Matsuno, Henry Tran, Hiroyuki Shimada
ABSTRACT
This study evaluated the utility of Phox2b in paediatric tumours. Previously, tyrosine hydroxylase (TH) was the most widely utilised sympathoadrenal marker specific for neural crest tumours with neuronal/neuroendocrine differentiation. However, its sensitivity is insufficient. Recently Phox2b has emerged as another specific marker for this entity. Phox2b immunohistochemistry (IHC) was performed on 159 paediatric tumours, including (group 1) 65 neural crest tumours with neuronal differentiation [peripheral neuroblastic tumours (pNT)]: 15 neuroblastoma undifferentiated (NB-UD), 10 NB poorly differentiated (NB-PD), 10 NB differentiating (NB-D), 10 ganglioneuroblastoma intermixed (GNBi), 10 GNB nodular (GNBn) and 10 ganglioneuroma (GN); (group 2) 23 neural crest tumours with neuroendocrine differentiation [pheochromocytoma/paraganglioma (PCC/PG)]; (group 3) 27 other neural crest tumours including one composite rhabdomyosarcoma/neuroblastoma; and (group 4) 44 non-neural crest tumours. TH IHC was performed on groups 1, 2 and 3. Phox2b was expressed diffusely in pNT (n = 65 of 65), strongly in NB-UD and NB-PD and with less intensity in NB-D, GNB and GN. Diffuse TH was seen in all NB-PD, NB-D, GNB and GN, but nine of 15 NB-UD and a nodule in GNBn did not express TH (n = 55 of 65). PCC/PG expressed diffuse Phox2b (n = 23 of 23) and diffuse TH, except for one tumour (n = 22 of 23). In composite rhabdomyosarcoma, TH was expressed only in neuroblastic cells and Phox2b was diffusely positive in neuroblastic cells and focally in rhabdomyosarcoma. All other tumours were negative for Phox2b (n = none of 44). Phox2b was a specific and sensitive marker for pNT and PCC/PG, especially useful for identifying NB-UD often lacking TH. Our study also presented a composite rhabdomyosarcoma/neuroblastoma of neural crest origin.
Product Description
Tyrosine Hydroxylase (LNC1) Mouse Monoclonal Antibody