MilliporeSigma
  • Oxicam-type nonsteroidal anti-inflammatory drugs enhance Agrobacterium-mediated transient transformation in plants.
Oxicam-type nonsteroidal anti-inflammatory drugs enhance Agrobacterium-mediated transient transformation in plants.
Plant biotechnology (Tokyo, Japan) (2022-11-10)
Seung-Won Choi, Kie Kumaishi, Reiko Motohashi, Harumi Enoki, Wiluk Chacuttayapong, Tadashi Takamizo, Hiroaki Saika, Masaki Endo, Tetsuya Yamada, Aya Hirose, Nobuya Koizuka, Seisuke Kimura, Yaichi Kawakatsu, Hiroyuki Koga, Emi Ito, Ken Shirasu, Yasunori Ichihashi
ABSTRACT
Agrobacterium-mediated transformation is a key innovation for plant breeding, and routinely used in basic researches and applied biology. However, the transformation efficiency is often the limiting factor of this technique. In this study, we discovered that oxicam-type nonsteroidal anti-inflammatory drugs, including tenoxicam (TNX), increase the efficiency of Agrobacterium-mediated transient transformation. TNX treatment increased the transformation efficiency of Agrobacterium-mediated transformation of Arabidopsis thaliana mature leaves by agroinfiltration. The increase of efficiency by TNX treatment was not observed in dde2/ein2/pad4/sid2 quadruple mutant, indicating that TNX inhibits the immune system mediated by jasmonic acid, ethylene, and salicylic acid against to Agrobacterium. We also found that TNX-treatment is applicable for the transient expression and subcellular localization analysis of fluorescent-tagged proteins in Arabidopsis leaf cells. In addition, we found that TNX increases the efficiency of Agrobacterium-mediated transient transformation of Jatropha. Given that treatment with oxicam compounds is a simple and cost effective method, our findings will provide a new option to overcome limitations associated with Agrobacterium-mediated transformation of various plant species.
Sigma-Aldrich
Tenoxicam, NSAID
Sigma-Aldrich
Piroxicam, ≥98% (TLC)