MilliporeSigma
  • KLF4-PFKFB3-driven glycolysis is essential for phenotypic switching of vascular smooth muscle cells.
KLF4-PFKFB3-driven glycolysis is essential for phenotypic switching of vascular smooth muscle cells.
Communications biology (2022-12-06)
Xinhua Zhang, Bin Zheng, Lingdan Zhao, Jiayi Shen, Zhan Yang, Yu Zhang, Ruirui Fan, Manli Zhang, Dong Ma, Lemin Zheng, Mingming Zhao, Huirong Liu, Jinkun Wen
ABSTRACT
Vascular smooth muscle cells (VSMCs) within atherosclerotic lesions undergo a phenotypic switching in a KLF4-dependent manner. Glycolysis plays important roles in transdifferentiation of somatic cells, however, it is unclear whether and how KLF4 mediates the link between glycolytic switch and VSMCs phenotypic transitions. Here, we show that KLF4 upregulation accompanies VSMCs phenotypic switching in atherosclerotic lesions. KLF4 enhances the metabolic switch to glycolysis through increasing PFKFB3 expression. Inhibiting glycolysis suppresses KLF4-induced VSMCs phenotypic switching, demonstrating that glycolytic shift is required for VSMCs phenotypic switching. Mechanistically, KLF4 upregulates expression of circCTDP1 and eEF1A2, both of which cooperatively promote PFKFB3 expression. TMAO induces glycolytic shift and VSMCs phenotypic switching by upregulating KLF4. Our study indicates that KLF4 mediates the link between glycolytic switch and VSMCs phenotypic transitions, suggesting that a previously unrecognized KLF4-eEF1A2/circCTDP1-PFKFB3 axis plays crucial roles in VSMCs phenotypic switching.
Sigma-Aldrich
Anti-LY6D antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.