MilliporeSigma
  • Diagnostic utility of WT-1 cytoplasmic stain in variety of vascular lesions.
Diagnostic utility of WT-1 cytoplasmic stain in variety of vascular lesions.
International journal of clinical and experimental pathology (2014-06-27)
Sarah K Galfione, Jae Y Ro, Alberto G Ayala, Yimin Ge
ABSTRACT
Vascular lesions are commonly encountered in routine pathologic practice and often pose diagnostic challenges owing to their morphologic diversity. Although WT-1 expression was reported in some vascular tumors, little is known about its staining patterns in a spectrum of vascular lesions from various locations. We examined WT-1 immunostain in 95 cases of vascular lesions including angiosarcomas (AS, 19 cases), hemangioendotheliomas (HE, 5), Kaposi's sarcomas (KS, 4), cavernous hemangiomas (CVH, 12), capillary hemangiomas (CPH, 7), pyogenic granulomas (PG, 4), lymphangiomas (LA, 4), hemangiopericytomas (HP, 5), glomus tumors (GT, 8), vascular malformation (VM, 13) and granulation tissue (GRT, 14). Strong WT-1 cytoplasmic stain was invariably observed in all cases of malignant and borderline vascular tumors including AS (19/19), KS (4/4) and HE (5/5). WT-1 was also consistently expressed in CPH (7/7), PG (4/4), and GRT (14/14), while it became weaker in VM (10/13) and often negative in CVH (2/12) and LA (0/4). WT1 stain was not demonstrated in HP (0/5) and rarely in GT (2/8). We conclude that consistent and diffuse WT-1 cytoplasmic stain in AS, HE and KS can be useful in distinguishing these tumors from poorly differentiated tumors with mimicking features. On the other hand, reliable WT-1 stain in CPH, PG and GRT may help in differential diagnosis with non-endothelial vascular tumors such as GT and HP. Recognizing the WT-1 cytoplasmic stain in a broad spectrum of benign and neoplastic tissues is critical in formulating appropriate immunohistochemical panels and avoiding misinterpretation of results.
Sigma-Aldrich
WT1 (6F-H2) Mouse Monoclonal Antibody
Pricing and availability is not currently available.