MilliporeSigma
  • Rapamycin and mTORC2 inhibition synergistically reduce contraction-stimulated muscle protein synthesis.
Rapamycin and mTORC2 inhibition synergistically reduce contraction-stimulated muscle protein synthesis.
The Journal of physiology (2020-09-08)
Riki Ogasawara, Jonas R Knudsen, Jingwen Li, Satoru Ato, Thomas E Jensen
ABSTRACT
Muscle contractions increase protein synthesis in a mechanistic target of rapamycin (mTOR)-dependent manner, yet it is unclear which/how mTOR complexes regulate muscle protein synthesis. We investigated the requirement of mTOR Complex 2 (mTORC2) in contraction-stimulated muscle protein synthesis. mTORC2 inhibition by muscle-specific Rictor knockout (Rictor mKO) did not prevent contraction-induced muscle protein synthesis. Rapamycin prevented contraction-induced muscle protein synthesis in Rictor mKO but not wild-type mice. Protein synthesis increases following muscle contractions. Previous studies have shown that inhibition of the mechanistic target of rapamycin complex 1 (mTORC1) suppresses the early but not late muscle protein synthesis response, while inhibition of both mTORC1 and mTORC2 abolishes the two effects. Therefore, we hypothesized that mTORC2 regulates muscle protein synthesis following muscle contractions. To test this, we investigated the effect of mTORC2 inhibition by mouse muscle-specific Rictor knockout (Rictor mKO) on muscle protein synthesis 3 h after contraction. The right gastrocnemius muscles of Rictor mKO and wild-type (WT) mice were isometrically contracted using percutaneous electrical stimulation, while the left gastrocnemius muscles served as controls. Vehicle or the mTORC1 inhibitor rapamycin (1.5 mg/kg) was injected intraperitoneally 1 h before contraction. Treatment of WT mice with rapamycin and Rictor mKO lowered protein synthesis in general, but the response to contractions was intact 3 h after contractions in both conditions. Rapamycin treatment in Rictor mKO mice prevented contraction-stimulated muscle protein synthesis. Notably, signalling traditionally associated with mTORC1 was increased by muscle contractions despite rapamycin treatment. In rapamycin-treated Rictor mKO mice, the same mTORC1 signalling was blocked following contractions. Our results indicate that although neither rapamycin-sensitive mTOR/mTORC1 nor mTORC2 is necessary for contraction-induced muscle protein synthesis, combined inhibition of rapamycin-sensitive mTOR/mTORC1 and mTORC2 synergistically inhibits contraction-induced muscle protein synthesis.
Sigma-Aldrich
Anti-SGK1 Antibody, clone 1G2.1, clone 1G2.1, from mouse
Sigma-Aldrich
Anti-Puromycin Antibody, clone 12D10, clone 12D10, from mouse