MilliporeSigma
  • Dose-response of the cultured bovine lens to butyl, methyl and propyl parabens.
Dose-response of the cultured bovine lens to butyl, methyl and propyl parabens.
International journal of cosmetic science (2008-05-21)
K Sivasegaran, L Ho, K L Moran, V Bantseev, J G Sivak
ABSTRACT
Pre-screening of cosmetic ingredients is vital for consumer safety. Previous in vivo techniques, such as the Draize test, have proved to be unreliable in predicting ocular irritancy and therefore there is a need for alternate testing methodologies. One such test is the scanning laser in vitro assay system which quantifies irritancy based on the focusing ability of the cultured bovine lens. In combination with confocal microscopy, a more thorough documentation of ocular irritancy can be achieved. This study investigates the response of cultured bovine lenses over time to butyl, methyl and propyl parabens, which are common antimicrobial agents found in cosmetic and ophthalmic products. The focusing ability of the lens was measured with an automated laser scanner over a period of 96 h. At 120 h post-treatment, the lenses were analysed by using a confocal laser scanning microscope to determine the characteristics of nuclei, and the morphology and distribution of mitochondria within the lenses. Irritancy to the three parabens was investigated at both an optical and cellular level. Each of the parabens was tested at 0.002% and 0.2%, where the 0.2% butyl paraben was found to be the most irritating.
Sigma-Aldrich
Gliadin from wheat