MilliporeSigma
  • A quantitative framework for the forward design of synthetic miRNA circuits.
A quantitative framework for the forward design of synthetic miRNA circuits.
Nature methods (2014-09-15)
Ryan J Bloom, Sally M Winkler, Christina D Smolke
ABSTRACT
Synthetic genetic circuits incorporating regulatory components based on RNA interference (RNAi) have been used in a variety of systems. A comprehensive understanding of the parameters that determine the relationship between microRNA (miRNA) and target expression levels is lacking. We describe a quantitative framework supporting the forward engineering of gene circuits that incorporate RNAi-based regulatory components in mammalian cells. We developed a model that captures the quantitative relationship between miRNA and target gene expression levels as a function of parameters, including mRNA half-life and miRNA target-site number. We extended the model to synthetic circuits that incorporate protein-responsive miRNA switches and designed an optimized miRNA-based protein concentration detector circuit that noninvasively measures small changes in the nuclear concentration of β-catenin owing to induction of the Wnt signaling pathway. Our results highlight the importance of methods for guiding the quantitative design of genetic circuits to achieve robust, reliable and predictable behaviors in mammalian cells.
Sigma-Aldrich
Sheep Anti-Rabbit IgG Antibody, HRP conjugate, 1 mg/mL, Chemicon®
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-HDAC1 Antibody, Upstate®, from rabbit
Pricing and availability is not currently available.