MilliporeSigma
Peptide substrates and inhibitors of the HIV-1 protease.
Biochemical and biophysical research communications (1989-03-15)
M L Moore, W M Bryan, S A Fakhoury, V W Magaard, W F Huffman, B D Dayton, T D Meek, L Hyland, G B Dreyer, B W Metcalf
ABSTRACT
Oligopeptides containing the consensus retroviral protease cleavage sequence Ser/Thr-X-Y-Tyr/Phe-Pro are substrates for purified recombinant HIV-1 protease with Km's in the millimolar range. The minimum sequence containing the consensus pentapeptide which serves as a good substrate is a heptapeptide spanning the P4-P3' residues. Substitution of reduced Phe-Pro or Tyr-Pro dipeptide isosteres or the statine analog 3-hydroxy-4-amino-5-phenylpentanoic acid for the scissile dipeptide afforded inhibitors of HIV-1 protease with Ki values in the micromolar range, three orders of magnitude better in affinity than the corresponding substrates. Inhibitors of HIV-1 protease may provide a novel and potentially useful therapeutic approach to the treatment of acquired immune deficiency syndrome (AIDS).