MilliporeSigma
  • 1,1,1,3,3,3-Hexafluoro-2-propanol (HFIP) as a novel and effective solvent to facilely prepare cyclodextrin-assembled materials.
1,1,1,3,3,3-Hexafluoro-2-propanol (HFIP) as a novel and effective solvent to facilely prepare cyclodextrin-assembled materials.
Chemical communications (Cambridge, England) (2014-10-07)
Toshiyuki Kida, Shin-ichiro Sato, Hiroaki Yoshida, Ayumi Teragaki, Mitsuru Akashi
ABSTRACT
We found that cyclodextrins (CDs) have a high solubility in 1,1,1,3,3,3-hexafluoro-2-propanol (HFIP). Evaporating HFIP from CD solutions on a glass plate gave crystalline solids composed of channel-type CD assemblies, and electrospinning with an HFIP solution of CDs fabricated CD microfibers.
Sigma-Aldrich
1,1,1,3,3,3-Hexafluoro-2-methyl-2-propanol, 97%