MilliporeSigma
Growth hormone prevents neuronal loss in the aged rat hippocampus.
Neurobiology of aging (2005-02-15)
Iñigo Azcoitia, Margarita Perez-Martin, Veronica Salazar, Carmen Castillo, Carmen Ariznavarreta, Luis M Garcia-Segura, Jesus A F Tresguerres
ABSTRACT
Decline of growth hormone (GH) with aging is associated to memory and cognitive alterations. In this study, the number of neurons in the hilus of the dentate gyrus has been assessed in male and female Wistar rats at 3, 6, 12, 14, 18, 22 and 24 months of age, using the optical fractionator method. Male rats had more neurons than females at all the ages studied. Significant neuronal loss was observed in both sexes between 22 and 24 months of age. In a second experiment, 22 month-old male and female rats were treated for 10 weeks with 2 mg/kg/day of GH or saline. At 24 months of age, animals treated with GH had more neurons in the hilus than animals treated with saline. These findings indicate that GH is neuroprotective in old animals and that its administration may ameliorate neuronal alterations associated to aging.
Product Description
Millipore
Rat/Mouse Growth Hormone ELISA, This Rat/Mouse Growth Hormone ELISA is used to measure & quantify Growth Hormone levels in Metabolism & Endocrine & Cell Signaling research.
Pricing and availability is not currently available.