MilliporeSigma
  • Advantages of Adult Mouse Dorsal Root Ganglia Explant Culture in Investigating Myelination in an Inherited Neuropathic Mice Model.
Advantages of Adult Mouse Dorsal Root Ganglia Explant Culture in Investigating Myelination in an Inherited Neuropathic Mice Model.
Methods and protocols (2022-07-28)
Yun Jeong Mo, Yu-Seon Kim, Minseok S Kim, Yun-Il Lee
ABSTRACT
A co-culture of neurons and Schwann cells has frequently been used to investigate myelin sheath formation. However, this approach is restricted to myelin-related diseases of the peripheral nervous system. This study introduces and compares an ex vivo model of adult-mouse-derived dorsal root ganglia (DRG) explant, with an in vitro co-culture of dissociated neurons from mouse embryo DRG and Schwann cells from a mouse sciatic nerve. The 2D co-culture has disadvantages of different mouse isolation for neurons and Schwann cells, animal number, culture duration, and the identification of disease model. However, 3D DRG explant neurons and myelination cells in Matrigel-coated culture are obtained from the same mouse, the culture period is shorter than that of 2D co-culture, and fewer animals are needed. In addition, it has simpler and shorter experimental steps than 2D co-culture. This culture system may prove advantageous in studies of biological functions and pathophysiological mechanisms of disease models, since it can reflect disease characteristics as traditional co-culture does. Therefore, it is suggested that a DRG explant culture is a scientifically, ethically, and economically more practical option than a co-culture system for studying myelin dynamics, myelin sheath formation, and demyelinating disease.
Sigma-Aldrich
Anti-Myelin Basic Protein Antibody, a.a. 82-87, culture supernatant, clone 12, Chemicon®