MilliporeSigma
  • Aflatoxins in Food and Feed: An Overview on Prevalence, Detection and Control Strategies.
Aflatoxins in Food and Feed: An Overview on Prevalence, Detection and Control Strategies.
Frontiers in microbiology (2019-10-23)
Dipendra K Mahato, Kyung Eun Lee, Madhu Kamle, Sheetal Devi, Krishna N Dewangan, Pradeep Kumar, Sang G Kang
ABSTRACT
Aflatoxins produced by the Aspergillus species are highly toxic, carcinogenic, and cause severe contamination to food sources, leading to serious health consequences. Contaminations by aflatoxins have been reported in food and feed, such as groundnuts, millet, sesame seeds, maize, wheat, rice, fig, spices and cocoa due to fungal infection during pre- and post-harvest conditions. Besides these food products, commercial products like peanut butter, cooking oil and cosmetics have also been reported to be contaminated by aflatoxins. Even a low concentration of aflatoxins is hazardous for human and livestock. The identification and quantification of aflatoxins in food and feed is a major challenge to guarantee food safety. Therefore, developing feasible, sensitive and robust analytical methods is paramount for the identification and quantification of aflatoxins present in low concentrations in food and feed. There are various chromatographic and sensor-based methods used for the detection of aflatoxins. The current review provides insight into the sources of contamination, occurrence, detection techniques, and masked mycotoxin, in addition to management strategies of aflatoxins to ensure food safety and security.
Sigma-Aldrich
Aflatoxin G2