MilliporeSigma
  • Synthesis of a CGRP Receptor Antagonist via an Asymmetric Synthesis of 3-Fluoro-4-aminopiperidine.
Synthesis of a CGRP Receptor Antagonist via an Asymmetric Synthesis of 3-Fluoro-4-aminopiperidine.
The Journal of organic chemistry (2019-05-28)
Carmela Molinaro, Eric M Phillips, Bangping Xiang, Erika Milczek, Michael Shevlin, Jaume Balsells, Scott Ceglia, Jiahui Chen, Lu Chen, Qinghao Chen, Zhongbo Fei, Scott Hoerrner, Ji Qi, Manuel de Lera Ruiz, Lushi Tan, Baoqiang Wan, Jingjun Yin
ABSTRACT
A practical and efficient enantioselective synthesis of the calcitonin gene-related peptide receptor antagonist 1 has been developed. The key structural component of the active pharmaceutical ingredient is a syn-1,2-amino-fluoropiperidine 4. Two approaches were developed to synthesize this important pharmacophore. Initially, Ru-catalyzed asymmetric hydrogenation of fluoride-substituted enamide 8 enabled the synthesis of sufficient quantities of compound 1 to support early preclinical studies. Subsequently, a novel, cost-effective route to this intermediate was developed utilizing a dynamic kinetic asymmetric transamination of ketone 9. This synthesis also features a robust Ullmann coupling to install a bis-aryl ether using a soluble Cu(I) catalyst. Finally, an enzymatic desymmetrization of meso-diester 7 was exploited for the construction of the γ-lactam moiety in 1.
Sigma-Aldrich
Bis[(tetrabutylammonium iodide)copper(I) iodide], 95%