MilliporeSigma
  • Activating CoOOH Porous Nanosheet Arrays by Partial Iron Substitution for Efficient Oxygen Evolution Reaction.
Activating CoOOH Porous Nanosheet Arrays by Partial Iron Substitution for Efficient Oxygen Evolution Reaction.
Angewandte Chemie (International ed. in English) (2018-02-09)
Sheng-Hua Ye, Zi-Xiao Shi, Jin-Xian Feng, Ye-Xiang Tong, Gao-Ren Li
ABSTRACT
Iron-substituted CoOOH porous nanosheet arrays grown on carbon fiber cloth (denoted as Fex Co1-x OOH PNSAs/CFC, 0≤x≤0.33) with 3D hierarchical structures are synthesized by in situ anodic oxidation of α-Co(OH)2 NSAs/CFC in solution of 0.01 m (NH4 )2 Fe(SO4 )2 . X-ray absorption fine spectra (XAFS) demonstrate that CoO6 octahedral structure in CoOOH can be partially substituted by FeO6 octahedrons during the transformation from α-Co(OH)2 to Fex Co1-x OOH, and this is confirmed for the first time in this study. The content of Fe in Fex Co1-x OOH, no more than 1/3 of Co, can be controlled by adjusting the in situ anodic oxidation time. Fe0.33 Co0.67 OOH PNSAs/CFC shows superior OER electrocatalytic performance, with a low overpotential of 266 mV at 10 mA cm-2 , small Tafel slope of 30 mV dec-1 , and high durability.
Sigma-Aldrich
Ammonium iron(II) sulfate hexahydrate, 99.997% trace metals basis