MilliporeSigma
  • Radiodynamic therapy with CsI(na)@MgO nanoparticles and 5-aminolevulinic acid.
Radiodynamic therapy with CsI(na)@MgO nanoparticles and 5-aminolevulinic acid.
Journal of nanobiotechnology (2022-07-17)
Fangchao Jiang, Chaebin Lee, Weizhong Zhang, Wen Jiang, Zhengwei Cao, Harrison Byron Chong, Wei Yang, Shuyue Zhan, Jianwen Li, Yong Teng, Zibo Li, Jin Xie
ABSTRACT
Radiodynamic therapy (RDT) holds the potential to overcome the shallow tissue penetration issue associated with conventional photodynamic therapy (PDT). To this end, complex and sometimes toxic scintillator-photosensitizer nanoconjugates are often used, posing barriers for large-scale manufacturing and regulatory approval. Herein, we report a streamlined RDT strategy based on CsI(Na)@MgO nanoparticles and 5-aminolevulinic acid (5-ALA). 5-ALA is a clinically approved photosensitizer, converted to protoporphyrin IX (PpIX) in cancer cells' mitochondria. CsI(Na)@MgO nanoparticles produce strong ~ 410 nm X-ray luminescence, which matches the Soret band of PpIX. We hypothesize that the CsI(Na)@MgO-and-5-ALA combination can mediate RDT wherein mitochondria-targeted PDT synergizes with DNA-targeted irradiation for efficient cancer cell killing. Because scintillator nanoparticles and photosensitizer are administered separately, the approach forgoes issues such as self-quenching or uncontrolled release of photosensitizers. When tested in vitro with 4T1 cells, the CsI(Na)@MgO and 5-ALA combination elevated radiation-induced reactive oxygen species (ROS), enhancing damages to mitochondria, DNA, and lipids, eventually reducing cell proliferation and clonogenicity. When tested in vivo in 4T1 models, RDT with the CsI(Na)@MgO and 5-ALA combination significantly improved tumor suppression and animal survival relative to radiation therapy (RT) alone. After treatment, the scintillator nanoparticles, made of low-toxic alkali and halide elements, were efficiently excreted, causing no detectable harm to the hosts. Our studies show that separately administering CsI(Na)@MgO nanoparticles and 5-ALA represents a safe and streamlined RDT approach with potential in clinical translation.
Sigma-Aldrich
Anti-phospho H2A.X (Ser139) Antibody, Alexa Fluor 647 Conjugate, from rabbit, ALEXA FLUOR 647