MilliporeSigma
  • Identification of oleoylethanolamide as an endogenous ligand for HIF-3α.
Identification of oleoylethanolamide as an endogenous ligand for HIF-3α.
Nature communications (2022-05-10)
Xiaotong Diao, Fei Ye, Meina Zhang, Xintong Ren, Xiaoxu Tian, Jingping Lu, Xiangnan Sun, Zeng Hou, Xiaoyu Chen, Fengwei Li, Jingjing Zhuang, Hong Ding, Chao Peng, Fraydoon Rastinejad, Cheng Luo, Dalei Wu
ABSTRACT
Hypoxia-inducible factors (HIFs) are α/β heterodimeric transcription factors modulating cellular responses to the low oxygen condition. Among three HIF-α isoforms, HIF-3α is the least studied to date. Here we show that oleoylethanolamide (OEA), a physiological lipid known to regulate food intake and metabolism, binds selectively to HIF-3α. Through crystallographic analysis of HIF-3 α/β heterodimer in both apo and OEA-bound forms, hydrogen-deuterium exchange mass spectrometry (HDX-MS), molecular dynamics (MD) simulations, and biochemical and cell-based assays, we unveil the molecular mechanism of OEA entry and binding to the PAS-B pocket of HIF-3α, and show that it leads to enhanced heterodimer stability and functional modulation of HIF-3. The identification of HIF-3α as a selective lipid sensor is consistent with recent human genetic findings linking HIF-3α with obesity, and demonstrates that endogenous metabolites can directly interact with HIF-α proteins to modulate their activities, potentially as a regulatory mechanism supplementary to the well-known oxygen-dependent HIF-α hydroxylation.
Sigma-Aldrich
N-Oleoylethanolamine, ~98% (TLC)