MilliporeSigma
  • REST Inactivation and Coexpression of ASCL1 and POU3F4 Are Necessary for the Complete Transformation of RB1/TP53-Inactivated Lung Adenocarcinoma into Neuroendocrine Carcinoma.
REST Inactivation and Coexpression of ASCL1 and POU3F4 Are Necessary for the Complete Transformation of RB1/TP53-Inactivated Lung Adenocarcinoma into Neuroendocrine Carcinoma.
The American journal of pathology (2022-04-04)
Meitetsu Masawa, Hanako Sato-Yazawa, Korehito Kashiwagi, Jun Ishii, Chie Miyata-Hiramatsu, Masami Iwamoto, Kakeru Kohno, Tadasuke Miyazawa, Masato Onozaki, Shuhei Noda, Yasuo Shimizu, Seiji Niho, Takuya Yazawa
ABSTRACT
Although recent reports have revealed the importance of the inactivation of both RB1 and TP53 in the transformation from lung adenocarcinoma into neuroendocrine carcinoma (NEC), the requirements for complete transformation into NEC have not been elucidated. To investigate alterations in the characteristics associated with the inactivation of RB1/TP53 and define the requirements for transformation into NEC cells, RB1/TP53 double-knockout A549 lung adenocarcinoma cells were established, and additional knockout of REST and transfection of ASCL1 and POU class 3 homeobox transcription factors (TFs) was conducted. More than 60 genes that are abundantly expressed in neural cells and several genes associated with epithelial-to-mesenchymal transition were up-regulated in RB1/TP53 double-knockout A549 cells. Although the expression of chromogranin A and synaptophysin was induced by additional knockout of REST (which mimics the status of most NECs), the expression of another neuroendocrine marker, CD56, and proneural TFs was not induced. However, coexpression of ASCL1 and POU3F4 in RB1/TP53/REST triple-knockout A549 cells induced the expression of not only CD56 but also other proneural TFs (NEUROD1 and insulinoma-associated 1) and induced NEC-like morphology. These findings suggest that the inactivation of RB1 and TP53 induces a state necessary for the transformation of lung adenocarcinoma into NEC and that further inactivation of REST and coexpression of ASCL1 and POU3F4 are the triggers for complete transformation into NEC.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-Brn-2 (POU3F2) Antibody, clone 8C4.2, clone 8C4.2, from mouse