MilliporeSigma
  • Endoplasmic reticulum stress is transmissible in vitro between cells of the central nervous system.
Endoplasmic reticulum stress is transmissible in vitro between cells of the central nervous system.
Journal of neurochemistry (2018-12-07)
Neil T Sprenkle, Anirudhya Lahiri, James W Simpkins, Gordon P Meares
ABSTRACT
Improper protein folding and trafficking are common pathological events in neurodegenerative diseases that result in the toxic accumulation of misfolded proteins within the lumen of the endoplasmic reticulum (ER). While low-level stimulation of the unfolded protein response (UPR) is protective, sustained UPR activation resulting from prolonged ER stress can promote neurotoxicity. The cell-autonomous mechanisms of the UPR have been extensively characterized. However, the cell-extrinsic role of the UPR under physiological and pathological states in the CNS remains to be elucidated. To begin to address this, we evaluated if transferring conditioned media between ER-stressed astrocytes and neurons could modulate their functional characteristics. Our results indicate that ER-stressed astrocytes and neurons secrete a molecule(s) with lipid characteristics which regulates both inflammatory and ER stress responses in other astrocytes, neurons, and microglia in vitro. Initial exposure to this stress factor(s) confers resistance against subsequent ER stress to neurons. However, persistent exposure to this unidentified mediator(s) suppresses the initial protective effect and becomes cytotoxic. Overall, these findings provide insight into the cell non-autonomous influence of ER stress on cells of the central nervous system. OPEN SCIENCE BADGES: This article has received a badge for *Open Materials* because it provided all relevant information to reproduce the study in the manuscript. The complete Open Science Disclosure form for this article can be found at the end of the article. More information about the Open Practices badges can be found at https://cos.io/our-services/open-science-badges/.