MilliporeSigma
  • Targeted degradation of transcription factors by TRAFTACs: TRAnscription Factor TArgeting Chimeras.
Targeted degradation of transcription factors by TRAFTACs: TRAnscription Factor TArgeting Chimeras.
Cell chemical biology (2021-04-10)
Kusal T G Samarasinghe, Saul Jaime-Figueroa, Michael Burgess, Dhanusha A Nalawansha, Katherine Dai, Zhenyi Hu, Adrian Bebenek, Scott A Holley, Craig M Crews
ABSTRACT
Many diseases, including cancer, stem from aberrant activation or overexpression of oncoproteins that are associated with multiple signaling pathways. Although proteins with catalytic activity can be successfully drugged, the majority of other protein families, such as transcription factors, remain intractable due to their lack of ligandable sites. In this study, we report the development of TRAnscription Factor TArgeting Chimeras (TRAFTACs) as a generalizable strategy for targeted transcription factor degradation. We show that TRAFTACs, which consist of a chimeric oligonucleotide that simultaneously binds to the transcription factor of interest (TOI) and to HaloTag-fused dCas9 protein, can induce degradation of the former via the proteasomal pathway. Application of TRAFTACs to two oncogenic TOIs, NF-κB and brachyury, suggests that TRAFTACs can be successfully employed for the targeted degradation of other DNA-binding proteins. Thus, TRAFTAC technology is potentially a generalizable strategy to induce degradation of other transcription factors both in vitro and in vivo.
Sigma-Aldrich
Rosetta 2(DE3) Competent Cells - Novagen, Novagen′s Rosetta 2 host strains are BL21 derivatives designed to enhance the expression of eukaryotic proteins that contain codons rarely used in E. coli.