MilliporeSigma
  • The squamous cell carcinoma cell line OM-1 retains both p75-dependent stratified epithelial progenitor potential and cancer stem cell properties.
The squamous cell carcinoma cell line OM-1 retains both p75-dependent stratified epithelial progenitor potential and cancer stem cell properties.
Biochemistry and biophysics reports (2021-05-28)
Ryo Uetsuki, Koichiro Higashikawa, Satoshi Okuda, Nao Yamakado, Fumi Ishida, Andra Rizqiawan, Shigehiro Ono, Masaaki Takechi, Kuniko Mizuta, Hideo Shigeishi, Nobuyuki Kamata, Kei Tobiume
ABSTRACT
The low-affinity nerve growth factor receptor p75 is a stratified epithelial stem/progenitor marker of human epithelia. We found OM-1, a human squamous cell carcinoma (SCC) cell line, showed distinct cells with p75 cluster, especially located at the center of a growing colony in a monolayer culture. A cell with p75 cluster was surrounded by cytokeratin 14- and cytokeratin 13-expressing cells that settled at the outer margin of the colony. OM-1 cells were also capable of forming tumor spheres in a cell suspension culture, an ability which was attenuated by the inhibition of p75-signaling. Intriguingly, we also found a p75-negative cell population from a growing culture of OM-1 that re-committed to become p75-clustering cells. These results indicated the possibility that SCC with epithelial multi-layering capacity can exploit the p75-dependent stratified epithelial progenitor property for the cancer stemness.
Sigma-Aldrich
p75NTR Signaling Inhibitor, Cell-permeable, TAT-Pep5, The p75NTR Signaling Inhibitor, Cell-permeable, TAT-Pep5 controls the biological activity of p75NTR. This small molecule/inhibitor is primarily used for Neuroscience applications.
Pricing and availability is not currently available.