MilliporeSigma
  • Telomeric 8-oxo-guanine drives rapid premature senescence in the absence of telomere shortening.
Telomeric 8-oxo-guanine drives rapid premature senescence in the absence of telomere shortening.
Nature structural & molecular biology (2022-07-01)
Ryan P Barnes, Mariarosaria de Rosa, Sanjana A Thosar, Ariana C Detwiler, Vera Roginskaya, Bennett Van Houten, Marcel P Bruchez, Jacob Stewart-Ornstein, Patricia L Opresko
ABSTRACT
Oxidative stress is a primary cause of cellular senescence and contributes to the etiology of numerous human diseases. Oxidative damage to telomeric DNA has been proposed to cause premature senescence by accelerating telomere shortening. Here, we tested this model directly using a precision chemoptogenetic tool to produce the common lesion 8-oxo-guanine (8oxoG) exclusively at telomeres in human fibroblasts and epithelial cells. A single induction of telomeric 8oxoG is sufficient to trigger multiple hallmarks of p53-dependent senescence. Telomeric 8oxoG activates ATM and ATR signaling, and enriches for markers of telomere dysfunction in replicating, but not quiescent cells. Acute 8oxoG production fails to shorten telomeres, but rather generates fragile sites and mitotic DNA synthesis at telomeres, indicative of impaired replication. Based on our results, we propose that oxidative stress promotes rapid senescence by producing oxidative base lesions that drive replication-dependent telomere fragility and dysfunction in the absence of shortening and shelterin loss.
Sigma-Aldrich
Benzonase® Nuclease, ultrapure, ≥250 units/μL, ≥99% (SDS-PAGE), recombinant, expressed in E. coli, buffered aqueous glycerol solution, ultrapure grade
Pricing and availability is not currently available.
Sigma-Aldrich
Cdk1 Inhibitor IV, RO-3306, RO-3306 is a cell-permeable, potent and ATP-competitive inhibitor of Cdk1 (Ki = 35 nM and 110 nM for Cdk1/B1 and Cdk1/A, respectively).
Pricing and availability is not currently available.