MilliporeSigma
  • A protocol to measure lysosomal Zn2+ release through a genetically encoded Zn2+ indicator.
A protocol to measure lysosomal Zn2+ release through a genetically encoded Zn2+ indicator.
STAR protocols (2022-06-17)
Mingxue Gu, Meiqin Hu, Taylor Minckley, Prateeksunder Pinchi, Haoxing Xu, Yan Qin, Wanlu Du
ABSTRACT
Intracellular vesicles such as lysosomes contain micromolar to millimolar concentrations of Zn2+, and disturbing lysosomal Zn2+ homeostasis via lysosomal Zn2+ release leads to mitochondria damage and consequent lytic cell death. Methods have been developed to image cellular Zn2+ dynamics. Here, we present a protocol using GZnP3, a genetically encoded fluorescent Zn2+ indicator, to assess lysosomal Zn2+ release in cultured cells by fluorescence microscopy imaging. For complete details on the use and execution of this protocol, please refer to Du et al. (2021) or Minckley et al. (2019).
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
INS-1 832/13 Rat Insulinoma Cell Line, INS-1 832/13 rat insulinoma cell line is a useful model for insulin secretion regulation and pancreatic islet beta-cell function studies.