MilliporeSigma
  • Niosomal Formulation of a Lipoyl-Carnosine Derivative Targeting TRPA1 Channels in Brain.
Niosomal Formulation of a Lipoyl-Carnosine Derivative Targeting TRPA1 Channels in Brain.
Pharmaceutics (2019-12-15)
Francesca Maestrelli, Elisa Landucci, Enrico De Luca, Giulia Nerli, Maria Camilla Bergonzi, Vieri Piazzini, Domenico E Pellegrini-Giampietro, Francesca Gullo, Andrea Becchetti, Francesco Tadini-Buoninsegni, Oscar Francesconi, Cristina Nativi
ABSTRACT
The transient receptor potential akyrin type-1 (TRPA1) is a non-selective cation channel playing a pivotal role in pain sensation and neurogenic inflammation. TRPA1 channels expressed in the central nervous system (CNS) have a critical role in the modulation of cortical spreading depression (CSD), which is a key pathophysiological basis of migraine pain. ADM_09 is a recently developed lipoic acid-based TRPA1 antagonist that is able to revert oxaliplatin-induced neuropathic pain and inflammatory trigeminal allodynia. In this context, aiming at developing drugs that are able to target TRPA1 channels in the CNS and promote an antioxidant effect, permeability across the blood-brain barrier (BBB) represents a central issue. Niosomes are nanovesicles that can be functionalized with specific ligands selectively recognized by transporters expressed on the BBB. In this work, the activity of ADM_09 on neocortex cultures was studied, and an efficient formulation to cross the BBB was developed with the aim of increasing the concentration of ADM_09 into the brain and selectively delivering it to the CNS rapidly after parenteral administration.
Sigma-Aldrich
Blood-Brain Barrier hCMEC/D3 Cell Line, The hCMEC/D3 BBB cell line has been extensively characterized for brain endothelial phenotype and is a model of human blood-brain barrier (BBB) function.