MilliporeSigma
  • Restoration of beta-galactosidase to Escherichia coli M15. Complementation studies.
Restoration of beta-galactosidase to Escherichia coli M15. Complementation studies.
The Biochemical journal (1977-09-01)
D V Marinkovic, J N Marinkovic
PMID411487
ABSTRACT
Carboxymethylated beta-galactosidase from Escherichia coli was dissociated at 100 degrees C to form carboxymethylated fragments A and B. The mol.wts. of carboxymethylated fragments A and B were determined by gel filtration to be 64300 and 22400 respectively. Sodium dodecyl sulphate/polyacrylamide-gel electrophoresis of carboxymethylated fragments A and B that had been pretreated with 2-mercaptoethanol and sodium dodecyl sulphate yielded mol.wts. of 64000 and 22100 respectively. Carboxymethylated fragments A and B had arginine as their C-terminal amino acid. When a crude extract of E. coli M15 was filtered through a column of Sepharose 6B, it was found that carboxymethylated fragment B could restore beta-galactosidase activity when added to fractions having mol.wts. estimated to be 123000, 262000 and 506000. These fractions are referred to as ;complementable fractions'. Similarly, it was found that carboxymethylated fragment A could restore enzyme activity to tractions having mol.wts. estimated to be 63000, 253000 and 506000. Estimates of the molecular weights of the beta-galactosidase activity obtained by restoration with carboxymethylated fragments A and B were made by filtering the active enzyme through another column of Sepharose 6B. The enzyme obtained by complementation with carboxymethylated fragment B, i.e. the complemented enzyme, had mol.wt. 525000, and that obtained with carboxymethylated fragment A had mol.wts. of 525000, 646000 and 2000000. The latter finding suggests that multiple forms of complemented beta-galactosidase can exist.
Sigma-Aldrich
HeLa + EGF Stimulated Cell Lysate in Mg2+ Lysis Buffer