MilliporeSigma
  • The role of cyclin D2 and p21/waf1 in human T-cell leukemia virus type 1 infected cells.
The role of cyclin D2 and p21/waf1 in human T-cell leukemia virus type 1 infected cells.
Retrovirology (2004-06-01)
Kylene Kehn, Longwen Deng, Cynthia de la Fuente, Katharine Strouss, Kaili Wu, Anil Maddukuri, Shanese Baylor, Robyn Rufner, Anne Pumfery, Maria Elena Bottazzi, Fatah Kashanchi
ABSTRACT
The human T-cell leukemia virus type 1 (HTLV-1) Tax protein indirectly influences transcriptional activation, signal transduction, cell cycle control, and apoptosis. The function of Tax primarily relies on protein-protein interactions. We have previously shown that Tax upregulates the cell cycle checkpoint proteins p21/waf1 and cyclin D2. Here we describe the consequences of upregulating these G1/S checkpoint regulators in HTLV-1 infected cells. To further decipher any physical and functional interactions between cyclin D2 and p21/waf1, we used a series of biochemical assays from HTLV-1 infected and uninfected cells. Immunoprecipitations from HTLV-1 infected cells showed p21/waf1 in a stable complex with cyclin D2/cdk4. This complex is active as it phosphorylates the Rb protein in kinase assays. Confocal fluorescent microscopy indicated that p21/waf1 and cyclin D2 colocalize in HTLV-1 infected, but not in uninfected cells. Furthermore, in vitro kinase assays using purified proteins demonstrated that the addition of p21/waf1 to cyclin D2/cdk4 increased the kinase activity of cdk4. These data suggest that the p21/cyclin D2/cdk4 complex is not an inhibitory complex and that p21/waf1 could potentially function as an assembly factor for the cyclin D2/cdk4 complex in HTLV-1 infected cells. A by-product of this assembly with cyclin D2/cdk4 is the sequestration of p21/waf1 away from the cyclin E/cdk2 complex, allowing this active cyclin-cdk complex to phosphorylate Rb pocket proteins efficiently and push cells through the G1/S checkpoint. These two distinct functional and physical activities of p21/waf1 suggest that RNA tumor viruses manipulate the G1/S checkpoint by deregulating cyclin and cdk complexes.