MilliporeSigma
  • Complex polyploid and hybrid species in an apomictic and sexual tropical forage grass group: genomic composition and evolution in Urochloa (Brachiaria) species.
Complex polyploid and hybrid species in an apomictic and sexual tropical forage grass group: genomic composition and evolution in Urochloa (Brachiaria) species.
Annals of botany (2021-12-08)
Paulina Tomaszewska, Maria S Vorontsova, Stephen A Renvoize, Sarah Z Ficinski, Joseph Tohme, Trude Schwarzacher, Valheria Castiblanco, José J de Vega, Rowan A C Mitchell, J S Heslop-Harrison
ABSTRACT
Diploid and polyploid Urochloa (including Brachiaria, Panicum and Megathyrsus species) C4 tropical forage grasses originating from Africa are important for food security and the environment​, often being planted in marginal lands worldwide. We aimed to characterize the nature of their genomes, the repetitive DNA, and the genome composition of polyploids, leading to a model of the evolutionary pathways within the group including many apomictic species. Some 362 forage grass accessions from international germplasm collections were studied, and ploidy determined using an optimized flow cytometry method. Whole-genome survey sequencing and molecular cytogenetic analysis were used to identify chromosomes and genomes in Urochloa accessions belonging to the 'brizantha' and 'humidicola' agamic complexes and U. maxima. Genome structures are complex and variable, with multiple ploidies and genome compositions within the species, and no clear geographical patterns. Sequence analysis of nine diploid and polyploid accessions enabled identification of abundant genome-specific repetitive DNA motifs​. In situ hybridization with a combination of repetitive DNA and genomic DNA probes, identified evolutionary divergence and allowed us to discriminate the different genomes present in polyploids. We suggest a new coherent nomenclature for the genomes present​. We develop a model of evolution at the whole-genome level in diploid and polyploid accessions showing processes of grass evolution. We support the retention of narrow species concepts for U. brizantha, U. decumbens, and U. ruziziensis, and do not consider diploids and polyploids of single species as cytotypes. The results and model will be valuable in making rational choices of parents for new hybrids, assist in use of the germplasm for breeding and selection of Urochloa with improved sustainability and agronomic potential, and will assist in measuring and conserving biodiversity in grasslands.
Sigma-Aldrich
Pectinase from Aspergillus niger, BioReagent, suitable for plant cell culture, aqueous glycerol solution, ≥5 units/mg protein (Lowry)
Sigma-Aldrich
Cellulase from Aspergillus niger, powder, ≥0.3 units/mg solid