MilliporeSigma
  • A Simple Method for Creating a High-Content Microscope for Imaging Multiplexed Tissue Microarrays.
A Simple Method for Creating a High-Content Microscope for Imaging Multiplexed Tissue Microarrays.
Current protocols (2021-04-07)
Shabnam Abtahi, Neal R Gliksman, John F Heneghan, Steven P Nilsen, Jeremy L Muhlich, Jay Copeland, Emil Rozbicki, Chris Allan, Pradeep K Dudeja, Jerrold R Turner
ABSTRACT
High-throughput, high-content imaging technologies and multiplex slide scanning have become widely used. Advantages of these approaches include the ability to archive digital copies of slides, review slides as teams using virtual microscopy software, and standardize analytical approaches. The cost and hardware and software inflexibility of dedicated slide scanning devices can, however, complicate implementation. Here, we describe a simple method that allows any microscope to be used for slide scanning. The only requirements are that the microscope be equipped with a motorized filter turret or wheels (for multi-channel fluorescence) and a motorized xyz stage. This example uses MetaMorph software, but the same principles can be used with any microscope control software that includes a few standard functions and allows programming of simple command routines, or journals. The series of journals that implement the method perform key functions, including assistance in defining an unlimited number of regions of interest (ROI) and imaging parameters. Fully automated acquisition is rapid, taking less than 3 hr to image fifty 2.5-mm ROIs in four channels. Following acquisition, images can be easily stitched and displayed using open-source or commercial image-processing and virtual microscope applications. © 2021 Wiley Periodicals LLC. Basic Protocol 1: Hardware and software configuration Basic Protocol 2: Create a preliminary scan Basic Protocol 3: Select, save, and position ROIs Basic Protocol 4: Determine and set autofocus parameters Basic Protocol 5: Acquire tiled images Basic Protocol 6: Review the scans Basic Protocol 7: Reimage ROIs as needed Basic Protocol 8: Stitch, stack, and assemble images Basic Protocol 9: Repeat scanning for multiplex immunostaining or background subtraction.
Sigma-Aldrich
Anti-SLC6A19 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution