MilliporeSigma
  • Expression and Purification of Recombinant Human Basic Fibroblast Growth Factor Fusion Proteins and Their Uses in Human Stem Cell Culture.
Expression and Purification of Recombinant Human Basic Fibroblast Growth Factor Fusion Proteins and Their Uses in Human Stem Cell Culture.
Journal of molecular microbiology and biotechnology (2015-11-20)
Sumeth Imsoonthornruksa, Kamthorn Pruksananonda, Rangsun Parnpai, Ruttachuk Rungsiwiwut, Mariena Ketudat-Cairns
ABSTRACT
To reduce the cost of cytokines and growth factors in stem cell research, a simple method for the production of soluble and biological active human basic fibroblast growth factor (hbFGF) fusion protein in Escherichia coli was established. Under optimal conditions, approximately 60-80 mg of >95% pure hbFGF fusion proteins (Trx-6xHis-hbFGF and 6xHis-hbFGF) were obtained from 1 liter of culture broth. The purified hbFGF proteins, both with and without the fusion tags, were biologically active, which was confirmed by their ability to stimulate proliferation of NIH3T3 cells. The fusion proteins also have the ability to support several culture passages of undifferentiated human embryonic stem cells and induce pluripotent stem cells. This paper describes a low-cost and uncomplicated method for the production and purification of biologically active hbFGF fusion proteins.
Sigma-Aldrich
Goat Anti-Mouse IgG Antibody, Cy3 conjugate, Species Adsorbed, 1.3 mg/mL, Chemicon®
Pricing and availability is not currently available.