MilliporeSigma
  • Baicalein Exerts Therapeutic Effects against Endotoxin-Induced Depression-like Behavior in Mice by Decreasing Inflammatory Cytokines and Increasing Brain-Derived Neurotrophic Factor Levels.
Baicalein Exerts Therapeutic Effects against Endotoxin-Induced Depression-like Behavior in Mice by Decreasing Inflammatory Cytokines and Increasing Brain-Derived Neurotrophic Factor Levels.
Antioxidants (Basel, Switzerland) (2022-05-29)
Hsin-Tzu Liu, Yu-Ning Lin, Ming-Cheng Tsai, Ya-Chi Wu, Ming-Chung Lee
ABSTRACT
Inflammation plays an important role in the pathophysiology of depression. This study aims to elucidate the antidepressant effect of baicalein, an anti-inflammatory component of a traditional Chinese herbal medicine (Scutellaria baicalensis), on lipopolysaccharide (LPS)-induced depression-like behavior in mice, and to investigate the underlying mechanisms. In vitro, baicalein exhibited antioxidant activity and protected macrophages from LPS-induced damage. The results of the tail suspension test and forced swimming test (tests for despair potential in mice) showed the antidepressant effect of baicalein on LPS-treated mice. It also substantially decreased the production of pro-inflammatory cytokines, including IL-6, TNF-α, MCP-1, and eotaxin, elicited by LPS in the plasma. Baicalein downregulated NF-κB-p65 and iNOS protein levels in the hippocampus, demonstrated its ability to mitigate neuroinflammation. Additionally, baicalein increased the levels of the mature brain-derived neurotrophic factor (mBDNF) in the hippocampus of LPS-treated mice, and elevated the ratio of mBDNF/proBDNF, which regulates neuronal survival and synaptic plasticity. Baicalein also promoted the expression of CREB, which plays a role in a variety of signaling pathways. In summary, the findings of this study demonstrate that the administration of baicalein can attenuate LPS-induced depression-like behavior by suppressing neuroinflammation and inflammation induced by the peripheral immune response.