MilliporeSigma
  • Downregulation of lncRNA FOXD2-AS1 Confers Radiosensitivity to Gastric Cancer Cells via miR-1913/SETD1A Axis.
Downregulation of lncRNA FOXD2-AS1 Confers Radiosensitivity to Gastric Cancer Cells via miR-1913/SETD1A Axis.
Cytogenetic and genome research (2022-03-31)
Fengying Guo, Ruixiang Guo, Licheng Zhang
ABSTRACT
Long noncoding RNA FOXD2 adjacent opposite strand RNA1 (FOXD2-AS1) plays an oncogenic role in various cancers, including gastric cancer (GC). However, the function of FOXD2-AS1 in regulating radiosensitivity of GC cells and its underlying molecular mechanisms have not been elucidated. This study aimed to figure out the potential mechanisms of FOXD2-AS1 in regulating GC cell radiosensitivity. RT-qPCR revealed upregulation of FOXD2-AS1 in GC cells exposed to radiation. Subcellular fractionation assay was used to localize FOXD2-AS1 in GC cells. Colony formation, MTT, EdU, and flow cytometry assays were performed to investigate the role of FOXD2-AS1 in regulating cell proliferation, cell cycle progression, and cell apoptosis. Western blotting was used to assess protein levels of apoptosis-associated markers and SET domain containing 1A (SETD1A). Homologous recombination reporter assay was conducted to explore the effect of FOXD2-AS1 on DNA damage repair. The downstream molecules of FOXD2-AS1 were identified with RNA pulldown, luciferase reporter, and RNA immunoprecipitation assays. The results showed that FOXD2-AS1 knockdown suppressed cell proliferation and cell cycle progression and promoted cell apoptosis and radiosensitivity of GC. FOXD2-AS1 could bind with miR-1913 in GC cells. In addition, miR-1913 targeted SETD1A, which was highly expressed in GC cells. Overexpression of SETD1A reversed FOXD2-AS1 silencing-induced effects on proliferation, apoptosis, and radiosensitivity of GC cells. In conclusion, knocking down FOXD2-AS1 enhances the radiosensitivity of GC cells by sponging miR-1913 to upregulate SETD1A expression.
Sigma-Aldrich
Anti-Ago 2 Antibody, Upstate®, from rabbit