MilliporeSigma
Dietary lignins are precursors of mammalian lignans in rats.
The Journal of nutrition (2004-01-06)
Aynun Nahar Begum, Catherine Nicolle, Isabelle Mila, Catherine Lapierre, Kazutane Nagano, Kazuhiko Fukushima, Satu-Maarit Heinonen, Herman Adlercreutz, Christian Rémésy, Augustin Scalbert
ABSTRACT
The mammalian lignans enterolactone (ENL) and enterodiol, commonly found in human plasma and urine, are phytoestrogens that may contribute to the prevention of breast cancer and coronary heart disease. They are formed by the conversion of dietary precursors such as secoisolariciresinol and matairesinol lignans by the colonic microflora. The identification of lignins, cell-wall polymers structurally related to lignans, as precursors of mammalian lignans is reported here for the first time. In study 1, rats were fed rye or wheat bran (15% diet) for 5 d. Untreated brans and brans extracted with solvents to remove lignans were compared. ENL was estimated in urine samples collected for 24 h by time-resolved fluoroimmunoassay. ENL urinary excretion was reduced from 18.6 to 5.3 nmol/d (n=8; P<0.001) when lignans were removed from rye bran and from 30.5 to 6.2 nmol/d (P<0.001) when they were removed from wheat bran. These results suggest that lignins, embedded in the cell wall and retained in the bran during solvent extraction, account for 26-32% of the ENL formed from cereal brans. In study 2, rats were fed a deuterated synthetic lignin (0.2% diet) together with wheat bran (15%) for 3 d. The detection of deuterated ENL by LC-tandem MS in urine (20 nmol/d) clearly confirms the conversion of lignin into mammalian lignans. More research is warranted to determine the bioavailability of lignins in the human diet.