MilliporeSigma
  • Simultaneous core 2 beta1-->6N-acetylglucosaminyltransferase up-regulation and sialyl-Le(X) expression during activation of human tonsillar B lymphocytes.
Simultaneous core 2 beta1-->6N-acetylglucosaminyltransferase up-regulation and sialyl-Le(X) expression during activation of human tonsillar B lymphocytes.
FEBS letters (1999-12-22)
M Nakamura, T Ishida, J Kikuchi, Y Furukawa, M Matsuda
ABSTRACT
We have investigated the regulation mechanism of the surface sialyl-Le(X) (sLe(X)) expression level in tonsillar B cells during activation. sLe(X) antigen became strongly positive after activation, while resting B cells were weakly positive. sLe(X) structures were mainly located on O-linked oligosaccharide chains of glycoprotein. Transcripts of FucT-VII and core 2 GlcNAc transferase (C2GnT) were up-regulated after activation, while those of ST3GalIV and beta1-->4GalT-I were expressed constitutively. However, the up-regulation of C2GnT was more dramatic than that of FucT-VII. These results suggest that sLe(X) expression level is regulated by C2GnT during tonsillar B cell activation.