MilliporeSigma
  • Interfering with mitochondrial dynamics sensitizes glioblastoma multiforme to temozolomide chemotherapy.
Interfering with mitochondrial dynamics sensitizes glioblastoma multiforme to temozolomide chemotherapy.
Journal of cellular and molecular medicine (2021-12-30)
Nan Wang, Renxuan Huang, Kunmeng Yang, Yichun He, Yufei Gao, Delu Dong
ABSTRACT
Glioblastoma multiforme (GBM) is a primary tumour of the central nervous system (CNS) that exhibits the highest degree of malignancy. Radiotherapy and chemotherapy are essential to prolong the survival time of patients. However, clinical work has demonstrated that sensitivity of GBM to chemotherapy decreases with time. The phenomenon of multi-drug resistance (MDR) reminds us that there may exist some fundamental mechanisms in the process of chemo-resistance. We tried to explore the mechanism of GBM chemo-resistance from the perspective of energy metabolism. First, we found that the oxidative phosphorylation (OXPHOS) level of SHG44 and U87 cells increased under TMZ treatment. In further studies, it was found that the expression of PINK1 and mitophagy flux downstream was downregulated in GBM cells, which were secondary to the upregulation of TP53 in tumour cells under TMZ treatment. At the same time, we examined the mitochondrial morphology in tumour cells and found that the size of mitochondria in tumour cells increased under the treatment of TMZ, which originated from the regulation of AMPK on the subcellular localization of Drp1 under the condition of unbalanced energy supply and demand in tumour cells. The accumulation of mitochondrial mass and the optimization of mitochondrial quality accounted for the increased oxidative phosphorylation, and interruption of the mitochondrial fusion process downregulated the efficiency of oxidative phosphorylation and sensitized GBM cells to TMZ, which was also confirmed in the in vivo experiment. What is more, interfering with this process is an innovative strategy to overcome the chemo-resistance of GBM cells.
Sigma-Aldrich
Anti-phospho-Ubiquitin (Ser65), from rabbit, purified by affinity chromatography