MilliporeSigma
  • Inhibitors of methionyl-tRNA synthetase have potent activity against Giardia intestinalis trophozoites.
Inhibitors of methionyl-tRNA synthetase have potent activity against Giardia intestinalis trophozoites.
Antimicrobial agents and chemotherapy (2015-09-02)
Ranae M Ranade, Zhongsheng Zhang, J Robert Gillespie, Sayaka Shibata, Christophe L M J Verlinde, Wim G J Hol, Erkang Fan, Frederick S Buckner
ABSTRACT
The methionyl-tRNA synthetase (MetRS) is a novel drug target for the protozoan pathogen Giardia intestinalis. This protist contains a single MetRS that is distinct from the human cytoplasmic MetRS. A panel of MetRS inhibitors was tested against recombinant Giardia MetRS, Giardia trophozoites, and mammalian cell lines. The best compounds inhibited trophozoite growth at 500 nM (metronidazole did so at ∼5,000 nM) and had low cytotoxicity against mammalian cells, indicating excellent potential for further development as anti-Giardia drugs.
Product Description
SAFC
Diamond Vitamin Tween® 80 Solution 40x, frozen liquid, suitable for cell culture