MilliporeSigma
Formononetin: Biological effects and uses - A review.
Food chemistry (2021-05-08)
Juliana Machado Dutra, Paula J P Espitia, Rejane Andrade Batista
ABSTRACT
Formononetin (FORM) is an isoflavone from the group of phytoestrogens that exhibits a broad spectrum of physiological effects beneficial to health through dependent and independent mechanisms of estrogen. This article aimed to present FORM main functions and future prospects for applications in different areas. Scientific publications and patents dated between 1998 and 2019 were analyzed. FORM has potential as an active compound of interest to product development for the industries of food, medicine, and cosmetics, among others. Moreover, in the medical area, this active compound has shown potential in the prevention and treatment of several diseases, including chronic ones, such as cancer, obesity, and neurodegenerative diseases.
Sigma-Aldrich
Formononetin, ≥99.0% (TLC)
Pricing and availability is not currently available.