MilliporeSigma
  • Peripheral vein infusion of autologous mesenchymal stem cells in Egyptian HCV-positive patients with end-stage liver disease.
Peripheral vein infusion of autologous mesenchymal stem cells in Egyptian HCV-positive patients with end-stage liver disease.
Stem cell research & therapy (2014-06-03)
Hosny Salama, Abdel-Rahman N Zekri, Eman Medhat, Shereen A Al Alim, Ola S Ahmed, Abeer A Bahnassy, Mai M Lotfy, Rasha Ahmed, Sherief Musa
ABSTRACT
We have assessed the utility of autologous mesenchymal stem cell (MSC) peripheral vein infusion as a possible therapeutic modality for patients with end-stage liver diseases. Forty patients with post-hepatitis C virus (HCV) end-stage liver disease were randomized into two groups: Group 1 (GI): 20 patients who received granulocyte colony-stimulating factor (G-CSF) for 5 days followed by autologous MSCs peripheral-vein infusion and group 2 (GII): 20 patients who received regular liver-supportive treatment only (control group). In MSC-infused patients (GI), 54% showed near normalization of liver enzymes and improvement in liver synthetic function. Significant changes were reported in albumin (P = 0.000), bilirubin (P = 0.002), increased international normalized ratio (INR) (P = 0.017), prothrombin concentration (P = 0.029) and alanine transaminase (ALT) levels (P = 0.029), with stabilization of clinical and biochemical status in 13% of cases. None of the patients in GII showed any significant improvement. Hepatic fibrosis was assessed in GI by detection of procollagen IIIC peptide level (PIIICP) and procollagen III N peptide level (PIIINP). The pretreatment values of s-PIIICP and s-PIIINP were 9.4 ± 4.2 and 440 ± 189, respectively, with a decrease to 8.1 ± 2.6 and 388 ± 102, respectively, 3 months after MSC therapy. However, the difference was statistically nonsignificant (P = 0.7). A significant correlation coefficient was reported after 3 months between the s-PIIINP and prothrombin concentration (P = -0.5) and between s-PIIICP and ascites (P = 0.550). First, autologous MSC infusion into a peripheral vein is as effective as the previously reported intrahepatic infusion. Second, MSCs have a supportive role in the treatment of end-stage liver disease, with satisfactory tolerability and beneficial effects on liver synthetic functions and hepatic fibrosis. Third, IV infusion of MSCs after G-CSF mobilization improves s-albumin within the first 2 weeks and prothrombin concentration and alanine Taransaminase after 1 month. According to the data from this current study and those previously reported by our group, we recommend further studies on patients' infusion with pure CD133 and CD34 followed by IV infusion of in vitro-differentiated MSCs within 1 week and another infusion after 3 months. ClinicalTrials.gov NCT01729221. Registered 17 November 2012.
Sigma-Aldrich
Gly-His-Lys acetate salt, BioXtra, γ-irradiated, suitable for cell culture