MilliporeSigma
  • Phenothiazine decorated carbazoles: effect of substitution pattern on the optical and electroluminescent characteristics.
Phenothiazine decorated carbazoles: effect of substitution pattern on the optical and electroluminescent characteristics.
The Journal of organic chemistry (2015-05-08)
Rajendra Kumar Konidena, K R Justin Thomas, Sudhir Kumar, Ya-Chi Wang, Chieh-Ju Li, Jwo-Huei Jou
ABSTRACT
A series of thienylphenothiazine decorated carbazoles were synthesized and characterized by optical, electrochemical, thermal, and theoretical investigations. Absorption spectra of the compounds are influenced by the substitution pattern and chromophore number density. Compounds containing 2,7-substitution exhibited red-shifted absorption, while the chromophore loading on the other positions led to the increment in molar extinction coefficients due to the increase in the chromophore density. Multiple substitutions resulted in twisting of chromophores and affected the conjugative delocalization of the π-electrons, which produced shorter wavelength absorption for the 2,3,6,7-tetrasubstituted derivative. Interestingly, the compounds exhibited excited-state solvatochromism attributable to the structural reorganization-induced electronic perturbations. The solvatochromic data are supportive of a general solvent effect, which is further confirmed by Lippert-Mataga correlation. End-capping with butterfly shaped phenothiazine restrained the formation of molecular aggregates in the solid state. All of the compounds displayed exceptional thermal stability attributable to the rigid carbazole building block. Solution processed OLED fabricated using the new materials as emitting dopants in 4,4'-bis(9H-carbazol-9-yl)biphenyl host exhibited bluish green electroluminescence.
Sigma-Aldrich
Phenothiazine, ≥98%
Pricing and availability is not currently available.