MilliporeSigma
  • Author Correction: Parvalbumin expression in oligodendrocyte-like CG4 cells causes a reduction in mitochondrial volume, attenuation in reactive oxygen species production and a decrease in cell processes' length and branching.
Author Correction: Parvalbumin expression in oligodendrocyte-like CG4 cells causes a reduction in mitochondrial volume, attenuation in reactive oxygen species production and a decrease in cell processes' length and branching.
Scientific reports (2020-07-23)
Lucia Lichvarova, Walter Blum, Beat Schwaller, Viktoria Szabolcsi
ABSTRACT
An amendment to this paper has been published and can be accessed via a link at the top of the paper.
Sigma-Aldrich
N1 Medium Supplement (100×), solution, sterile-filtered, suitable for cell culture
Pricing and availability is not currently available.