MilliporeSigma
  • Survival genes expression analysis following ionizing radiation to LiCl treated KG1a cells.
Survival genes expression analysis following ionizing radiation to LiCl treated KG1a cells.
International journal of radiation biology (2020-01-28)
Yogesh Kumar Verma, Ajay Kumar Singh, Gangenahalli Ugraiah Gurudutta
ABSTRACT
Purpose: Lithium chloride (LiCl) is clinically used for manic disorders. Its role has been shown in improving cell survival by decreasing Bax and p53 expression and increasing Bcl-2 concentration in the cell. This potential of LiCl is responsible for reducing irradiated cell death. In this study, we have explored the role of LiCl as a radioprotectant affecting survival genes.Materials and methods: To find out the cellular response upon LiCl pretreatment to radiation-exposed KG1a cells; viability, clonogenic assay and microarray studies were performed. This was followed by the detection of transcription factor binding motif in coregulated genes. These results were confirmed by reverse transcription-polymerase chain reaction (RT-PCR) and chromatin immunoprecipitation (CHIP).Results: LiCl improved irradiated KG1a cell survival and its clonogenicity at 2 mM concentration (clinically used). Microarray data analysis showed differential expression of cell-protecting genes playing an important role in apoptosis, cell cycle, adhesion and inflammation, etc. The coregulation analysis revealed genes involved in bile acid biosynthesis were also affected by LiCl treatment, these genes are likely to be responsible for radiation-induced gastrointestinal (GI) syndrome through bile production.Conclusions: This is the first study with respect to global genetic expression upon LiCl treatment to radiation-exposed cells. Our results suggest considering repurposing of LiCl as a protective agent for radiation injury.
Sigma-Aldrich
Iscove′s Modified Dulbecco′s Medium, With L-glutamine and 25 mM HEPES, without sodium bicarbonate, powder, suitable for cell culture
Pricing and availability is not currently available.