MilliporeSigma
  • Resolving the chemical identity of H2SO4 derived anions on Pt(111) electrodes: they're sulfate.
Resolving the chemical identity of H2SO4 derived anions on Pt(111) electrodes: they're sulfate.
Physical chemistry chemical physics : PCCP (2019-08-23)
Igor Ying Zhang, Gregor Zwaschka, Zhenhua Wang, Martin Wolf, R Kramer Campen, Yujin Tong
ABSTRACT
Understanding how electrolyte composition controls electrocatalytic reactions requires molecular-level insight into electrode/electrolyte interaction. Perhaps the most basic aspect of this interaction, the speciation of the interfacial ion, is often controversial for even relatively simple systems. For example, for Pt(111) in 0.5 M H2SO4 it has long been debated whether the adsorbed anion is SO42-, HSO4- or an H3O+SO42- ion pair. Here we apply interface-specific vibrational sum frequency (VSF) spectroscopy and theory to this problem and perform an isotope exchange study: we collect VSF spectra of Pt(111) in H2SO4(H2O) and D2SO4(D2O) as a function of bias and show that at all potentials they are identical. This is the most direct spectroscopic evidence to date that SO42- is the dominant adsorbate, despite the fact that at 0.5 M H2SO4 bulk solution is dominated by HSO4-. This approach is based on the unique selection rule of the VSF spectroscopy and thus offers a new way of accessing general electrode/electrolyte interaction in electrocatalysis.
Sigma-Aldrich
Sulfuric acid-d2 solution, 96-98 wt. % in D2O, 99.5 atom % D
Pricing and availability is not currently available.