MilliporeSigma
Three functional facets of calbindin D-28k.
Frontiers in molecular neuroscience (2012-03-22)
Hartmut Schmidt
ABSTRACT
Many neurons of the vertebrate central nervous system (CNS) express the Ca(2+) binding protein calbindin D-28k (CB), including important projection neurons like cerebellar Purkinje cells but also neocortical interneurons. CB has moderate cytoplasmic mobility and comprises at least four EF-hands that function in Ca(2+) binding with rapid to intermediate kinetics and affinity. Classically it was viewed as a pure Ca(2+) buffer important for neuronal survival. This view was extended by showing that CB is a critical determinant in the control of synaptic Ca(2+) dynamics, presumably with strong impact on plasticity and information processing. Already 30 years ago, in vitro studies suggested that CB could have an additional Ca(2+) sensor function, like its prominent acquaintance calmodulin (CaM). More recent work substantiated this hypothesis, revealing direct CB interactions with several target proteins. Different from a classical sensor, however, CB appears to interact with its targets both, in its Ca(2+)-loaded and Ca(2+)-free forms. Finally, CB has been shown to be involved in buffered transport of Ca(2+), in neurons but also in kidney. Thus, CB serves a threefold function as buffer, transporter and likely as a non-canonical sensor.