MilliporeSigma
  • N-arachidonoyl dopamine inhibits epithelial-mesenchymal transition of breast cancer cells through ERK signaling and decreasing the cellular cholesterol.
N-arachidonoyl dopamine inhibits epithelial-mesenchymal transition of breast cancer cells through ERK signaling and decreasing the cellular cholesterol.
Journal of biochemical and molecular toxicology (2021-01-05)
Shreetama Bandyopadhayaya, Mikhail G Akimov, Ranjeet Verma, Ankit Sharma, Divya Sharma, Gopal C Kundu, Natalia M Gretskaya, Vladimir V Bezuglov, Chandi C Mandal
ABSTRACT
N-acyl dopamines (NADAs) are bioactive lipids of the endovanilloid family with known cytotoxicity for the cancer cells; however, the available data on the participation of the endovanilloids in epithelial-mesenchymal transition (EMT) and cancer stemness are controversial. This study unveils the inhibitory role of N-arachidonoyl dopamine (AA-DA), a typical representative of the NADA family, in breast cancer cell migration, EMT, and stemness. AA-DA treatment also led to a decrease in cholesterol biosynthesis gene expressions, and addition of exogenous cholesterol reverted these AA-DA-mediated inhibitory effects. Notably, AA-DA treatment inhibited the key regulatory gene of the cholesterol biosynthesis pathway, sterol regulatory element-binding protein 1 (SREBP1), with concurrent repression of the endoplasmic reticulum kinase 1/2 (ERK1/2) pathway. Furthermore, U0126, an ERK inhibitor, inhibited SREBP1 and decreased cellular cholesterol level, unwinding the molecular mechanism behind AA-DA-mediated anticancer activity. Thus, we, for the first time, revealed that AA-DA counteracts breast cancer EMT via inhibition of ERK signaling and cholesterol content.
Sigma-Aldrich
Anti-SREBP1 Antibody, clone 2121, clone 2121, Upstate®, from mouse