MilliporeSigma
  • Properties of TAN-67, a nonpeptidic delta-opioid receptor agonist, at cloned human delta- and mu-opioid receptors.
Properties of TAN-67, a nonpeptidic delta-opioid receptor agonist, at cloned human delta- and mu-opioid receptors.
European journal of pharmacology (1995-10-15)
R J Knapp, R Landsman, S Waite, E Malatynska, E Varga, W Haq, V J Hruby, W R Roeske, H Nagase, H I Yamamura
ABSTRACT
2-methyl-4a alpha-(3-hydroxyphenyl)-1,2,3,4,4a,5,12,12a alpha-octahydro-quinolino[2,3,30g]isoquinoline (TAN-67) is a nonpeptidic delta-opioid receptor agonist. This report describes its receptor binding affinity and agonist potency at human and mouse delta and mu-opioid receptors. The binding affinities of TAN-67 and the cyclic enkephalin analog, (D-Pen2, 4'-Cl-Phe4, D-Pen5]enkephalin (pCl-DPDPE) were measured by radioligand binding inhibition studies at mouse and human variants of the delta and mu-opioid receptor using [3H]Naltrindole and [3H]D-Phe-Cys-Tyr-D-Trp-Orn-Thr -Pen-Thr-NH2, respectively. TAN-67 showed high binding affinity (Ki = 0.647 nM) at the human delta-opioid receptor and high delta-opioid receptor binding selectivity ( > 1000-fold) relative to the human mu-opioid receptor. TAN-67 also showed high potency (EC50 = 1.72 nM) for the inhibition of forskolin-stimulated cAMP accumulation at human delta-opioid receptors expressed by intact Chinese hamster ovary cells but low potency (EC50 = 1520 nM) at human mu-opioid receptors expressed by intact B82 mouse fibroblast cells. The results show that TAN-67 has similar binding affinities, selectivity and potencies as pCl-DPDPE at human delta and mu-opioid receptors. These results combined with the nonpeptidic structure of TAN-67 suggest that this compound has therapeutic potential as a delta-opioid receptor agonist.