MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • The intersection of genetic and chemical genomic screens identifies GSK-3α as a target in human acute myeloid leukemia.
The intersection of genetic and chemical genomic screens identifies GSK-3α as a target in human acute myeloid leukemia.
The Journal of clinical investigation (2012-02-14)
Versha Banerji, Stacey M Frumm, Kenneth N Ross, Loretta S Li, Anna C Schinzel, Cynthia K Hahn, Rose M Kakoza, Kwan T Chow, Linda Ross, Gabriela Alexe, Nicola Tolliday, Haig Inguilizian, Ilene Galinsky, Richard M Stone, Daniel J DeAngelo, Giovanni Roti, Jon C Aster, William C Hahn, Andrew L Kung, Kimberly Stegmaier
ABSTRACT
Acute myeloid leukemia (AML) is the most common form of acute leukemia in adults. Long-term survival of patients with AML has changed little over the past decade, necessitating the identification and validation of new AML targets. Integration of genomic approaches with small-molecule and genetically based high-throughput screening holds the promise of improved discovery of candidate targets for cancer therapy. Here, we identified a role for glycogen synthase kinase 3α (GSK-3α) in AML by performing 2 independent small-molecule library screens and an shRNA screen for perturbations that induced a differentiation expression signature in AML cells. GSK-3 is a serine-threonine kinase involved in diverse cellular processes, including differentiation, signal transduction, cell cycle regulation, and proliferation. We demonstrated that specific loss of GSK-3α induced differentiation in AML by multiple measurements, including induction of gene expression signatures, morphological changes, and cell surface markers consistent with myeloid maturation. GSK-3α-specific suppression also led to impaired growth and proliferation in vitro, induction of apoptosis, loss of colony formation in methylcellulose, and anti-AML activity in vivo. Although the role of GSK-3β has been well studied in cancer development, these studies support a role for GSK-3α in AML.
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)