MilliporeSigma
  • Enhancement of immunogenicity of a therapeutic cervical cancer DNA-based vaccine by co-application of sequence-optimized genetic adjuvants.
Enhancement of immunogenicity of a therapeutic cervical cancer DNA-based vaccine by co-application of sequence-optimized genetic adjuvants.
International journal of cancer (2009-04-10)
Peter Ohlschläger, Michael Quetting, Gerardo Alvarez, Matthias Dürst, Lutz Gissmann, Andreas M Kaufmann
ABSTRACT
Treatment of patients with cervical cancer by conventional methods (mainly surgery, but also radiotherapy and chemotherapy) results in a significant loss in quality of life. A therapeutic DNA vaccine directed to tumor-specific antigens of the human papilloma virus (HPV) could be an attractive treatment option. We have developed a nontransforming HPV-16 E7-based DNA vaccine containing all putative T cell epitopes (HPV-16 E7SH). DNA vaccines, however, are less immunogenic than protein- or peptide-based vaccines in larger animals and humans. In this study, we have investigated an adjuvant gene support of the HPV-16 E7SH therapeutic cervical cancer vaccine. DNA encoded cytokines (IL-2, IL-12, GM-CSF, IFN-gamma) and the chemokine MIP1-alpha were co-applied either simultaneously or at different time points pre- or post-E7SH vaccination. In addition, sequence-optimized adjuvant genes were compared to wild type genes. Three combinations investigated lead to an enhanced IFN-gamma response of the induced T cells in mice. Interestingly, IFN-gamma secretion of splenocytes did not strictly correlate with tumor response in tumor regression experiments. Gene-encoded MIP-1alpha applied 5 days prior to E7SH-immunization combined with IFN-gamma or IL-12 (3 days) or IL-2 (5 days) postimmunization lead to a significantly enhanced tumor response that was clearly associated with granzyme B secretion and target cells lysis. Our results suggest that a conditioning application and combination with adjuvant genes may be a promising strategy to enhance synergistically immune responses by DNA immunization for the treatment of cervical cancer.
Sigma-Aldrich
Interferon-γ Protein, Recombinant mouse, The Interferon-gamma protein (or IFN-gamma protein) is a regulatory protein produced by activated NK cells & CD4+TCRalpha/beta+, CD8+TCRalpha/beta+ & TCRgamma/delta+ T cells.